missing translation for 'onlineSavingsMsg'
Learn More

7-Epi-10-oxo-docetaxel, MedChemExpress

Product Code. 16489657
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489657 5 mg 5mg
16451629 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489657 Supplier Medchem Express Supplier No. HY16675
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 7-Epi-10-oxo-docetaxel (Docetaxel Impurity 2) is a impurity of docetaxel detected by high performance liquid chromatography (HPLC).

7-Epi-10-oxo-docetaxel (Docetaxel Impurity 2) is a impurity of docetaxel detected by high performance liquid chromatography (HPLC).

Specifications

Chemical Name or Material 7-Epi-10-oxo-docetaxel
CAS 162784-72-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C43H51NO14
Quantity 5 mg
Synonym Docetaxel Impurity 2
SMILES CC(O[C@@]1([C@](C[C@H]2O)([H])OC1)[C@@]([C@@]2(C3=O)C)([H])[C@@H]([C@@](C4(C)C)(C[C@H](OC([C@H](O)[C@H](C5=CC=CC=C5)NC(OC(C)(C)C)=O)=O)C(C)=C4C3=O)O)OC(C6=CC=CC=C6)=O)=O
Molecular Weight (g/mol) 805.86
Formula Weight 805.86
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.