missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 7-Diethylamino-4-methylcoumarin, 99%

Product Code. p-108093 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10395740

Brand: Thermo Scientific Chemicals 114050050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-44-1 | C14H17NO2 | 231.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-44-1
Molecular Formula C14H17NO2
Molecular Weight (g/mol) 231.29
MDL Number MFCD00006864
InChI Key AFYCEAFSNDLKSX-UHFFFAOYSA-N
Synonym 7-diethylamino-4-methylcoumarin, coumarin 1, 7-diethylamino-4-methyl-2h-chromen-2-one, coumarin 47, coumarin 460, 4-methyl-7-diethylamino coumarin, blancophor aw, blancophor ffg, 4-methyl-7-diethylaminocoumarin, hakkol p
PubChem CID 7050
ChEBI CHEBI:51938
IUPAC Name 7-(diethylamino)-4-methylchromen-2-one
SMILES CCN(CC)C1=CC2=C(C=C1)C(=CC(=O)O2)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 70°C to 73°C
Color Beige to Purple
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 5 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in aqueous acid solutions,soluble in nearly all common organic solvents,slightly soluble in aliphatic hydrocarbons
Formula Weight 231.29
Percent Purity 99%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material 7-Diethylamino-4-methylcoumarin, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 7-Diethylamino-4-methylcoumarin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 7-Diethylamino-4-methylcoumarin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.