missing translation for 'onlineSavingsMsg'
Learn More

7-Deazahypoxanthine, 97%

Product Code. 15401809
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15401809 5 g 5g
15491799 1 g 1g
15411809 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15401809 Supplier Thermo Scientific Alfa Aesar Supplier No. H64537.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3680-71-5 | C6H5N3O | 135.13 g/mol

It is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Moisture and light sensitive. Store at -20°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3680-71-5
Molecular Formula C6H5N3O
Molecular Weight (g/mol) 135.13
MDL Number MFCD08445809
InChI Key FBMZEITWVNHWJW-UHFFFAOYSA-N
Synonym 7h-pyrrolo 2,3-d pyrimidin-4-ol, pyrrolo 2,3-d pyrimidin-4-ol, 7-deazahypoxanthine, 3h-pyrrolo 2,3-d pyrimidin-4 7h-one, 4-hydroxypyrrolo 2,3-d pyrimidine, 4-hydroxypyrrolopyrimidine, 1h-pyrrolo 2,3-d pyrimidin-4 7h-one, 3,7-dihydro-4h-pyrrolo 2,3-d pyrimidin-4-one, pyrrolo 2,3-d pyrimidin-4-one
PubChem CID 96194
IUPAC Name 1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
SMILES O=C1N=CNC2=C1C=CN2

Specifications

Melting Point 345°C to 350°C
Color Brown
Quantity 5 g
Beilstein 607869
Sensitivity Light and moisture sensitive
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 135.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 7-Deazahypoxanthine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.