missing translation for 'onlineSavingsMsg'
Learn More

7-Deazaguanine, 97%, Thermo Scientific™

Product Code. 15400858
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400858 250 mg 250mg
15490848 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400858 Supplier Thermo Scientific Alfa Aesar Supplier No. H54595.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethidium bromide efficiently intercalates into synthetic duplex DNAs with 7-deazaguanine in place of guanine, but shows none of the fluorescence enhancement usually observed upon intercalation. 7-Deazaadenine-duplexes show about 70% of the expected fluorescence enhancement.

Solubility
Soluble in N,N-Dimethylformamide, Dimethyl sulfoxide.

Notes
Store at room temperature. Keep away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7355-55-7
Molecular Formula C6H6N4O
Molecular Weight (g/mol) 150.14
MDL Number MFCD09033259
InChI Key OLAFFPNXVJANFR-UHFFFAOYSA-N
Synonym 7-deazaguanine, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, unii-gpl8t5zo3m, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, gpl8t5zo3m, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one
PubChem CID 96253
IUPAC Name 2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
SMILES NC1=NC(=O)C2=C(NC=C2)N1

Specifications

Quantity 250 mg
Beilstein 512680
Solubility Information Soluble in N,N-Dimethylformamide,Dimethyl sulfoxide.
Formula Weight 150.14
Percent Purity 97%
Chemical Name or Material 7-Deazaguanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.