missing translation for 'onlineSavingsMsg'
Learn More

7-Chlorokynurenic acid sodium salt, MedChemExpress

Product Code. 16443178
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16443178

Brand: Medchem Express HY100811A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 7-Chlorokynurenic acid sodium salt (7-CKA sodium salt) is a potent and selective antagonist of the glycine B coagonist site of the N-methyl-D-aspartate (NMDA) receptor (IC50=0.56 μM). 7-Chlorokynurenic acid sodium salt is also a potent inhibitor of the reuptake of glutamate into synaptic vesicles with a Ki of 0.59 μM. 7-Chlorokynurenic acid sodium salt has potent antinociceptive actions after neuraxial delivery.

7-Chlorokynurenic acid sodium salt (7-CKA sodium salt) is a potent and selective antagonist of the glycine B coagonist site of the N-methyl-D-aspartate (NMDA) receptor (IC50=0.56 μM). 7-Chlorokynurenic acid sodium salt is also a potent inhibitor of the reuptake of glutamate into synaptic vesicles with a Ki of 0.59 μM. 7-Chlorokynurenic acid sodium salt has potent antinociceptive actions after neuraxial delivery.

Specifications

Chemical Name or Material 7-Chlorokynurenic acid sodium salt
Physical Form Solid
CAS 1263094-00-3
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H5ClNNaO3
For Use With (Application) Neuroscience-Neurodegeneration
Synonym 7-CKA sodium salt
Solubility Information H2O : 50 mg/mL (203.59 mM; Need ultrasonic) ∣DMSO : ≥ 33.67 mg/mL (137.10 mM)
SMILES O=C(C1=NC2=CC(Cl)=CC=C2C(O)=C1)O[Na]
Molecular Weight (g/mol) 245.59
Formula Weight 245.59
Percent Purity 99.32%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.