missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Chlorokynurenic acid sodium salt, MedChemExpress
Description
7-Chlorokynurenic acid sodium salt (7-CKA sodium salt) is a potent and selective antagonist of the glycine B coagonist site of the N-methyl-D-aspartate (NMDA) receptor (IC50=0.56 μM). 7-Chlorokynurenic acid sodium salt is also a potent inhibitor of the reuptake of glutamate into synaptic vesicles with a Ki of 0.59 μM. 7-Chlorokynurenic acid sodium salt has potent antinociceptive actions after neuraxial delivery.
Specifications
Specifications
| Chemical Name or Material | 7-Chlorokynurenic acid sodium salt |
| Physical Form | Solid |
| CAS | 1263094-00-3 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C10H5ClNNaO3 |
| For Use With (Application) | Neuroscience-Neurodegeneration |
| Synonym | 7-CKA sodium salt |
| Solubility Information | H2O : 50 mg/mL (203.59 mM; Need ultrasonic) ∣DMSO : ≥ 33.67 mg/mL (137.10 mM) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?