missing translation for 'onlineSavingsMsg'
Learn More

7-Chloroisatin, 97%

Product Code. p-7052744
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15441908

Brand: Thermo Scientific Alfa Aesar H55594.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7477-63-6 | C8H4ClNO2 | 181.575 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7477-63-6
Molecular Formula C8H4ClNO2
Molecular Weight (g/mol) 181.575
MDL Number MFCD00022796
InChI Key MPLXQMMMGDYXIT-UHFFFAOYSA-N
PubChem CID 344135
IUPAC Name 7-chloro-1H-indole-2,3-dione
SMILES C1=CC2=C(C(=C1)Cl)NC(=O)C2=O

Specifications

Melting Point 187°C to 191°C
Color Orange
Quantity 1 g
Synonym 7-chloroisatin, 7-chloroindoline-2,3-dione, 7-chloro-2,3-dihydro-1h-indole-2,3-dione, 7-chloro-2,3-indolinedione, 1h-indole-2,3-dione, 7-chloro, 1h-indole-2,3-dione,7-chloro, 7-chloro-1h-benzo d azolidine-2,3-dione, 7-chlorisatin, 7-chloro isatin, pubchem15537
Formula Weight 181.58
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 7-Chloroisatin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.