missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-6-nitro-4(3H)-quinazolinone, 97%, Thermo Scientific™

Product Code. 15254854 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15254854 5 g Glass bottle 5g
15264854 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254854 Supplier Thermo Scientific Acros Supplier No. 450790050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 53449-14-2
Molecular Formula C8H4ClN3O3
Molecular Weight (g/mol) 225.59
InChI Key URDYTQYZXZKBQT-UHFFFAOYSA-N
Synonym 7-chloro-6-nitroquinazolin-4 3h-one, 7-chloro-4-hydroxy-6-nitroquinazoline, 7-chloro-6-nitro-4-hydroxyquinazoline, 6-nitro-7-chloro-4-hydroxyquinazoline, 7-chloro-6-nitroquinazolin-4-ol, 7-chloro-6-nitroquinazolin-4-one, 7-chloro-6-nitro-quinazolin-4-one, 7-chloro-6-nitro-3h-quinazolin-4-one, 4 1h-quinazolinone, 7-chloro-6-nitro, 7-chloro-6-nitro-3,4-dihydroquinazolin-4-one
PubChem CID 12441237
IUPAC Name 7-chloro-6-nitro-1H-quinazolin-4-one
SMILES C1=C2C(=CC(=C1[N+](=O)[O-])Cl)NC=NC2=O

Specifications

Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: slightly soluble in dmf
Formula Weight 225.59
Percent Purity 97%
Chemical Name or Material 7-Chloro-6-nitro-4(3H)-quinazolinone
compliance-icons
Product Identifier
  • 7-Chloro-6-nitroquinazolin-4-ol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.