missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-6-fluoro-1-(2,4-difluorophenyl)-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic Acid Ethyl Ester, TRC

Product Code. 30360580
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30360580 1 g 1g
30355009 10 g -
30352728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30360580 Supplier Toronto Research Chemicals Supplier No. TRCC3663501G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100491-29-0
Molecular Formula C17H10ClF3N2O3
Molecular Weight (g/mol) 382.72
Synonym 7-Chloro-1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic Acid Ethyl Ester, Ethyl 1-(2,4-Difluorophenyl)-7-chloro-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylate
IUPAC Name ethyl 7-chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylate
SMILES CCOC(=O)C1=CN(c2ccc(F)cc2F)c3nc(Cl)c(F)cc3C1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H10ClF3N2O3/c1-2-26-17(25)10-7-23(13-4-3-8(19)5-11(13)20)16-9(14(10)24)6-12(21)15(18)22-16/h3-7H,2H2,1H3
Formula Weight 382.0332
Chemical Name or Material 7-Chloro-6-fluoro-1-(2,4-difluorophenyl)-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.