missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-6-azaindole, 97%, Thermo Scientific™

Catalog Number 11492190
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
357263-41-3
C7H5ClN2
152.58
MFCD03839905
HOHKYYCVFMEBGG-UHFFFAOYSA-N
11435048
7-chloro-1H-pyrrolo[2,3-c]pyridine
ClC1=C2NC=CC2=CC=N1
This item is not returnable. View return policy
357263-41-3
C7H5ClN2
152.58
MFCD03839905
HOHKYYCVFMEBGG-UHFFFAOYSA-N
11435048
7-chloro-1H-pyrrolo[2,3-c]pyridine
ClC1=C2NC=CC2=CC=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Synonym 7-chloro-1h-pyrrolo 2,3-c pyridine, 7-chloro-6-azaindole, 1h-pyrrolo 2,3-c pyridine, 7-chloro, acmc-1aep0, 7-chloropyrrolo 2,3-c pyridine, 1h-pyrrolo 2,3-c pyridine,7-chloro, 7-chloranyl-1h-pyrrolo 2,3-c pyridine, 7-chloro-1h-pyrrolo 2 pound not3-c pyridine
Formula Weight 152.58
Percent Purity 97%
Chemical Name or Material 7-Chloro-6-azaindole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'