missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Chloro-4-hydroxyquinoline, 99%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Melting Point | 276.0°C to 279.0°C |
Assay Percent Range | 98.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 25 g |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 179.61 |
Percent Purity | 99% |
Chemical Name or Material | 7-Chloro-4-hydroxyquinoline |
Safety and Handling
Product Identifier
- 7-Chloro-4-hydroxyquinoline
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'