missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-3-methyl-4H-1,2,4-benzothiadiazine 1-Oxide, TRC

Product Code. 30351757
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351757 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351757 Supplier Toronto Research Chemicals Supplier No. TRCC6055755MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 7-Chloro-3-methyl-4H-1,2,4-benzothiadiazine 1-Oxide
CAS 31365-76-1
Molecular Formula C8H7ClN2OS
Quantity 5 mg
Synonym 4H-1,2,4-Benzothiadiazine, 7-chloro-3-methyl-, 1-oxide (8CI, ACI), 7-Chloro-3-methyl-4H-1,2,4-benzothiadiazine 1-oxide
InChI Formula InChI=1S/C8H7ClN2OS/c1-5-10-7-3-2-6(9)4-8(7)13(12)11-5/h2-4H,1H3,(H,10,11)
SMILES CC1=Nc2ccc(Cl)cc2S(=O)N1
IUPAC Name 7-chloro-3-methyl-2H-1λ^{4},2,4-benzothiadiazine 1-oxide
Molecular Weight (g/mol) 214.67
Formula Weight 213.9968
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.