missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-1,2,3,4-tetrahydroisoquinoline, 95%, Thermo Scientific Chemicals

Product Code. 15480519
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82771-60-6
Molecular Formula C9H10ClN
Molecular Weight (g/mol) 167.64
MDL Number MFCD05861543
InChI Key BLRKQLICTKRDDZ-UHFFFAOYSA-N
Synonym 7-chloro-1,2,3,4-tetrahydro-isoquinoline, isoquinoline, 7-chloro-1,2,3,4-tetrahydro, 7-chhloro-1,2,3,4-tetrahydroisoquinoline, 7-chloranyl-1,2,3,4-tetrahydroisoquinoline, isoquinoline,7-chloro-1,2,3,4-tetrahydro
PubChem CID 12595069
IUPAC Name 7-chloro-1,2,3,4-tetrahydroisoquinoline
SMILES ClC1=CC2=C(CCNC2)C=C1

Specifications

Quantity 1 g
Formula Weight 167.64
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 7-Chloro-1,2,3,4-tetrahydroisoquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'