missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-1,2,3,4-tetrahydro-1-(2-methyl-4-nitrobenzoyl)-5H-1-benzazepin-5-one, TRC

Product Code. 30367062
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367062 250 mg 250mg
30366731 50 mg 50mg
30359435 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30367062 Supplier Toronto Research Chemicals Supplier No. TRCC366123250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137982-91-3
Molecular Formula C18H15ClN2O4
Molecular Weight (g/mol) 358.78
Synonym 1-(2-Methyl-4-nitrobenzoyl)-7-chloro-1,2,3,4-tetrahydrobenzo[b]azepin-5-one, 7-Chloro-1-(2-methyl-4-nitrobenzoyl)-5-oxo-2,3,4,5+tetrahydro-1H-1-benzazepine, 7-Chloro-1-(2-methyl-4-nitrobenzoyl)-5-oxo-2,3,4,5-tetrahydro-1H-1-benzazepine
IUPAC Name 7-chloro-1-(2-methyl-4-nitrobenzoyl)-3,4-dihydro-2H-1-benzazepin-5-one
SMILES Cc1cc(ccc1C(=O)N2CCCC(=O)c3cc(Cl)ccc23)[N+](=O)[O-]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C18H15ClN2O4/c1-11-9-13(21(24)25)5-6-14(11)18(23)20-8-2-3-17(22)15-10-12(19)4-7-16(15)20/h4-7,9-10H,2-3,8H2,1H3
Formula Weight 358.072
Chemical Name or Material 7-Chloro-1,2,3,4-tetrahydro-1-(2-methyl-4-nitrobenzoyl)-5H-1-benzazepin-5-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.