missing translation for 'onlineSavingsMsg'
Learn More

7-Bromobenzo[b]thiophene, 97%, Thermo Scientific™

Product Code. 10307364
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10307364 1 g Amber Glass Bottle 1g
10255434 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10307364 Supplier Thermo Scientific Maybridge Supplier No. CC29410DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1423-61-6
Molecular Formula C8H5BrS
Molecular Weight (g/mol) 213.09
MDL Number MFCD06657729
InChI Key NOICDPBEDNMHQK-UHFFFAOYSA-N
Synonym 7-bromobenzo b thiophene, 7-bromobenzothiophene, 7-bromo-benzo b thiophene, benzo b thiophene, 7-bromo, zlchem 1192, 7-bromobenzo-b thiophene, benzo b thiophene,7-bromo
PubChem CID 12045538
IUPAC Name 7-bromo-1-benzothiophene
SMILES BrC1=C2SC=CC2=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 213.1
Percent Purity 97%
Chemical Name or Material 7-Bromobenzo[b]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.