missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoacetyl-3,4-dihydro-1,5-benzodioxepin, 97%

Product Code. 11302399
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11302399

Brand: Thermo Scientific Alfa Aesar H27314.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35970-34-4 | C11H11BrO3 | 271.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35970-34-4
Molecular Formula C11H11BrO3
Molecular Weight (g/mol) 271.11
MDL Number MFCD00218462
InChI Key YJSZSLGVTLSCRU-UHFFFAOYSA-N
Synonym 2-bromo-1-3,4-dihydro-2h-1,5-benzodioxepin-7-yl ethan-1-one, 2-bromo-1-3,4-dihydro-2h-1,5-benzodioxepin-7-yl ethanone, 7-bromoacetyl-3,4-dihydro-1,5-benzodioxepin, 2-bromo-1-3,4-dihydro-2h-benzo b 1,4 dioxepin-7-yl ethanone, 2-bromo-1-3,4-dihydro-2h-benzo b 1,4 dioxepin-7-yl ethan-1-one, ethanone, 2-bromo-1-3,4-dihydro-2h-1,5-benzodioxepin-7-yl, acmc-1ahn7, 3,4-trimethylenedioxy phenacyl bromide, 1-2h,3h,4h-benzo b 1,4-dioxepan-7-yl-2-bromoethan-1-one, 1-3,4-dihydro-2h-1,5-benzodioxepin-7-yl-2-bromoethanone
PubChem CID 2776389
IUPAC Name 2-bromo-1-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)ethanone
SMILES C1COC2=C(C=C(C=C2)C(=O)CBr)OC1

Specifications

Melting Point 74°C to 78°C
Quantity 1 g
UN Number UN1759
Formula Weight 271.11
Percent Purity 97%
Chemical Name or Material 7-Bromoacetyl-3,4-dihydro-1,5-benzodioxepin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.