missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-5-methylbenzo[b]furan, 97%

Product Code. 15427287
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427287 250 mg 250mg
15417287 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427287 Supplier Thermo Scientific Alfa Aesar Supplier No. H34347.MD
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 35700-48-2 | C9H7BrO | 211.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 35700-48-2
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.058
MDL Number MFCD11877828
InChI Key TZUMDTURHSPEAY-UHFFFAOYSA-N
Synonym 7-bromo-5-methylbenzofuran, 7-bromo-5-methylbenzo b furan, 7-bromo-5-methyl-benzofuran, benzofuran, 7-bromo-5-methyl
PubChem CID 21071801
IUPAC Name 7-bromo-5-methyl-1-benzofuran
SMILES CC1=CC(=C2C(=C1)C=CO2)Br

Tekniske data

Quantity 250 mg
Formula Weight 211.06
Percent Purity 97%
Chemical Name or Material 7-Bromo-5-methylbenzo[b]furan

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.