missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-5-methyl-1-benzofuran, 95%, Thermo Scientific™

Product Code. 10027739
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10027739 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10027739 Supplier Thermo Scientific Maybridge Supplier No. MAY00316DA
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 35700-48-2
Assay Percent Range 95%
Molecular Formula C9H7BrO
Quantity 1 g
Synonym 7-bromo-5-methylbenzofuran, 7-bromo-5-methylbenzo b furan, 7-bromo-5-methyl-benzofuran, benzofuran, 7-bromo-5-methyl
InChI Key TZUMDTURHSPEAY-UHFFFAOYSA-N
SMILES CC1=CC(=C2C(=C1)C=CO2)Br
IUPAC Name 7-bromo-5-methyl-1-benzofuran
Molecular Weight (g/mol) 211.058
PubChem CID 21071801
Formula Weight 211.06
Chemical Name or Material 7-Bromo-5-methyl-1-benzofuran
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.