missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-4-methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine, 95%, Thermo Scientific™

Product Code. 10758825
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10758825

Brand: Thermo Scientific Maybridge CC62010DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 910037-14-8
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C8H9BrN2O
MDL Number MFCD09025911
Quantity 1 g
Synonym 7-bromo-4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine, 7-bromo-4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazine, pubchem17432, 7-bromo-4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazine
InChI Key KPGNYZHIKPSZMS-UHFFFAOYSA-N
SMILES CN1CCOC2=CC(=CN=C21)Br
IUPAC Name 7-bromo-4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazine
Molecular Weight (g/mol) 229.077
PubChem CID 24229649
Formula Weight 229.08
Percent Purity 95%
Chemical Name or Material 7-Bromo-4-methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.