missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-3,4-dihydro-1(2H)-naphthalenone, 97%, Thermo Scientific™

Product Code. 10351685
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10351685 25 g 25g
10631435 1 g 1g
10330543 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10351685 Supplier Thermo Scientific Maybridge Supplier No. MO01275EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 32281-97-3
Molecular Formula C10H9BrO
Molecular Weight (g/mol) 225.085
MDL Number MFCD02179287
InChI Key YGVDCGFUUUJCDF-UHFFFAOYSA-N
Synonym 7-bromo-1-tetralone, 7-bromo-3,4-dihydronaphthalen-1 2h-one, 7-bromo-3,4-dihydro-1 2h-naphthalenone, 7-bromotetralone, 7-bromo-alpha-tetralone, 7-bromotetralin-1-one, 7-bromo-1,2,3,4-tetrahydronaphthalen-1-one, 7-bromo-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone, 7-bromo-3,4-dihydro, 7-bromo tetralone
PubChem CID 252731
IUPAC Name 7-bromo-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=C(C=C2)Br)C(=O)C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 225.08
Percent Purity 97%
Chemical Name or Material 7-Bromo-3, 4-dihydro-1(2H)-naphthalenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.