missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10255654
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10255654 1 g Amber Glass Bottle 1g
11376805 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10255654 Supplier Thermo Scientific Maybridge Supplier No. CC58101DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 879896-63-6
Molecular Formula C7H5BrO4S
Molecular Weight (g/mol) 265.077
MDL Number MFCD08690304
InChI Key KIURUSHZGLZLSQ-UHFFFAOYSA-N
Synonym 7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxylic acid, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxine-5-carboxylic acid, 7-bromo-2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carboxylic acid, thieno 3,4-b-1,4-dioxin-5-carboxylicacid, 7-bromo-2,3-dihydro
PubChem CID 18525889
IUPAC Name 5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-7-carboxylic acid
SMILES C1COC2=C(SC(=C2O1)C(=O)O)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 265.08
Percent Purity 97%
Chemical Name or Material 7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.