missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-methyl-1H-indole, 97%, Thermo Scientific™

Product Code. 10434635
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10434635

Brand: Thermo Scientific Maybridge CC66910DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 280752-68-3
Molecular Formula C9H8BrN
Molecular Weight (g/mol) 210.07
MDL Number MFCD09879960
InChI Key CALOMHQMSHLUJX-UHFFFAOYSA-N
Synonym 7-bromo-1-methyl-1h-indole, 1h-indole,7-bromo-1-methyl, 1h-indole, 7-bromo-1-methyl, 1-methyl-7-bromoindole, 1-methyl-7-bromo-1h-indole
PubChem CID 21950069
IUPAC Name 7-bromo-1-methylindole
SMILES CN1C=CC2=CC=CC(Br)=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 210.07
Percent Purity 97%
Chemical Name or Material 7-Bromo-1-methyl-1H-indole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.