missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-heptene, 97%

Product Code. 15451189
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15451189 100 g 100g
15431189 5 g 5g
15441189 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451189 Supplier Thermo Scientific Alfa Aesar Supplier No. H64127.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4117-09-3 | C7H13Br | 177.09 g/mol

It is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4117-09-3
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.09
MDL Number MFCD01631137
InChI Key GNYDYUQVALBGGZ-UHFFFAOYSA-N
IUPAC Name 7-bromohept-1-ene

Specifications

Color Colorless to Yellow
Density 1.162 g/mL
Boiling Point 27°C (1 mmHg)
Flash Point 58°C (136°F)
Linear Formula Br(CH2)5CH=CH2
Refractive Index 1.466
Quantity 100 g
UN Number UN1993
Beilstein 1740861
Synonym 2-amino-1,3,5-triazine, amino-1,3,5-triazine, 2-amino-s-triazine, 1,3,5-triazine-2-ylamine, chembl89436, amino-s-triazine, s-triazine-2-amine, pubchem14498, 1,3,5 triazin-2-ylamine
Solubility Information Slightly soluble in water.
SMILES BrCCCCCC=C
PubChem CID 20077
Formula Weight 177.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 7-Bromo-1-heptene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.