missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-heptene, 97%

Product Code. 15431189
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
15431189 5 g 5g
15441189 25 g 25g
15451189 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15431189 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor H64127.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 4117-09-3 | C7H13Br | 177.09 g/mol

It is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 4117-09-3
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.09
MDL Number MFCD01631137
InChI Key GNYDYUQVALBGGZ-UHFFFAOYSA-N
IUPAC Name 7-bromohept-1-ene

Especificaciones

Color Colorless to Yellow
Density 1.162 g/mL
Boiling Point 27°C (1 mmHg)
Flash Point 58°C (136°F)
Linear Formula Br(CH2)5CH=CH2
Refractive Index 1.466
Quantity 5 g
UN Number UN1993
Beilstein 1740861
Synonym 7-bromo-1-heptene, 1-heptene, 7-bromo, bromo-7 heptene-1, 7-bromo-heptene, 7-bromo-l-heptene, 1-bromo-6-heptene, acmc-2097en, ksc235k6r
Solubility Information Slightly soluble in water.
SMILES C=CCCCCCBr
PubChem CID 524450
Formula Weight 177.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 7-Bromo-1-heptene
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • 7-Bromo-1-heptene
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contain: 7-Bromo-1-heptene

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.