missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-heptene, 97%

Product Code. 15441189
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15441189

Brand: Thermo Scientific Alfa Aesar H64127.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4117-09-3 | C7H13Br | 177.09 g/mol

It is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4117-09-3
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.09
MDL Number MFCD01631137
InChI Key GNYDYUQVALBGGZ-UHFFFAOYSA-N
IUPAC Name 7-bromohept-1-ene

Specifications

Color Colorless to Yellow
Density 1.162 g/mL
Boiling Point 27°C (1 mmHg)
Flash Point 58°C (136°F)
Linear Formula Br(CH2)5CH=CH2
Refractive Index 1.466
Quantity 25 g
UN Number UN1993
Beilstein 1740861
Synonym 7-bromo-1-heptene, 1-heptene, 7-bromo, bromo-7 heptene-1, 7-bromo-heptene, 7-bromo-l-heptene, 1-bromo-6-heptene, acmc-2097en, ksc235k6r
Solubility Information Slightly soluble in water.
SMILES C=CCCCCCBr
PubChem CID 524450
Formula Weight 177.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 7-Bromo-1-heptene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 7-Bromo-1-heptene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 7-Bromo-1-heptene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.