missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-heptanol, 96%

Product Code. 15440968
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440968 1 g 1g
15450968 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440968 Supplier Thermo Scientific Alfa Aesar Supplier No. H54762.03
  • ONLINE EXCLUSIVE PRICE
    £59.11
    £48.65 / 1g
    Save £10.46
    17% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10160-24-4 | C7H15BrO | 195.1 g/mol

7-Bromo-1-heptanol was used in the synthesis of (9Z,12E)-[1-13C]-octadeca-9,12-dienoic acid and(9Z,12Z,15E)-[1-13C]-octadeca-9,12,15-trienoic acid. It can be used to prepare N-substituted [(carboxymethyl)phenoxy]ethyl benzoxazinones as PPARγ agonists and antidiabetic (type 2) agents. It can also be used to prepare thiochroman and chroman derivatives as pure antiestrogens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromo-1-heptanol was used in the synthesis of (9Z,12E)-[1-13C]-octadeca-9,12-dienoic acid and(9Z,12Z,15E)-[1-13C]-octadeca-9,12,15-trienoic acid. It can be used to prepare N-substituted [(carboxymethyl)phenoxy]ethyl benzoxazinones as PPARγ agonists and antidiabetic (type 2) agents. It can also be used to prepare thiochroman and chroman derivatives as pure antiestrogens.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10160-24-4
Molecular Formula C7H15BrO
Molecular Weight (g/mol) 195.1
MDL Number MFCD00041708
InChI Key MMXRRNUXCHUHOE-UHFFFAOYSA-N
Synonym 7-bromo-1-heptanol, 1-heptanol, 7-bromo, 7-bromoheptanol, 7-bromoheptane-1-ol
PubChem CID 66284
IUPAC Name 7-bromoheptan-1-ol
SMILES C(CCCO)CCCBr

Specifications

Density 1.269
Boiling Point 111°C to 112°C (4 mmHg)
Flash Point 113°C (235°F)
Linear Formula Br(CH2)7OH
Refractive Index 1.482
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 195.1
Percent Purity 96%
Chemical Name or Material 7-Bromo-1-heptanol
compliance-icons
Product Identifier
  • 7-Bromo-1-heptanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 7-Bromo-1-heptanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.