missing translation for 'onlineSavingsMsg'
Learn More

7-(Beta-Hydroxypropyl)theophylline, TRC

Product Code. 30402344
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402344 10 mg 10mg
30397578 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402344 Supplier Toronto Research Chemicals Supplier No. TRCH95264010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-00-9
Molecular Formula C10 H14 N4 O3
Molecular Weight (g/mol) 238.24
Synonym Proxyphylline, 7-[(2RS)-2-Hydroxypropyl]-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione, Diprophylline Imp. D (EP)
IUPAC Name 7-(2-hydroxypropyl)-1,3-dimethylpurine-2,6-dione
SMILES CC(O)Cn1cnc2N(C)C(=O)N(C)C(=O)c12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H14N4O3/c1-6(15)4-14-5-11-8-7(14)9(16)13(3)10(17)12(8)2/h5-6,15H,4H2,1-3H3
Formula Weight 238.1066
Chemical Name or Material Proxyphylline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.