missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-beta-Amino-7-alpha-methoxy-3-(1-methyl-5-tetrazolylthio)methyl-3-cephem-4-carboxylic Acid Diphenylmethyl Ester, TRC
Specifications
Specifications
| CAS | 56610-72-1 |
| Molecular Formula | C24H24N6O4S2 |
| Quantity | 1 g |
| Synonym | (6R,7S)-7-Amino-7-methoxy-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Diphenylmethyl Ester, 7-MAC, (6R-cis)-7-Amino-7-methoxy-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Diphenylmethyl Ester, Diphenylmethyl 7β-Amino-7α-methoxy-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-3-cephem-4-carboxylate; |
| InChI Formula | InChI=1S/C24H24N6O4S2/c1-29-23(26-27-28-29)36-14-17-13-35-22-24(25,33-2)21(32)30(22)18(17)20(31)34-19(15-9-5-3-6-10-15)16-11-7-4-8-12-16/h3-12,19,22H,13-14,25H2,1-2H3/t22-,24+/m1/s1 |
| SMILES | CN1C(=NN=N1)SCC2=C(C(=O)OC(C3=CC=CC=C3)C4=CC=CC=C4)N5C(=O)[C@]([C@H]5SC2)(N)OC |
| IUPAC Name | benzhydryl (6R,7S)-7-amino-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
| Molecular Weight (g/mol) | 524.62 |
| Chemical Name or Material | 7beta-Amino-7alpha-methoxy-3-(1-methyl-5-tetrazolylthio)methyl-3-cephem-4-carboxylic Acid Diphenylmethyl Ester |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?