missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 7-Benzyloxyindole-3-carboxaldehyde, 97%

Product Code. 15493827
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493827 250 mg 250mg
11462140 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493827 Supplier Alfa Aesar™ Supplier No. H28777.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 92855-65-7
Molecular Formula C16H13NO2
Molecular Weight (g/mol) 251.285
MDL Number MFCD00055979
InChI Key RNARWQXZOKWYHY-UHFFFAOYSA-N
Synonym 7-benzyloxyindole-3-carbaldehyde, 7-benzyloxy-3-formylindole, 7-benzyloxy-1h-indole-3-carbaldehyde, 7-benzyloxyindole-3-carboxaldehyde, pubchem14131, 7-phenylmethoxy indole-3-carbaldehyde, 7-benzyloxy-1h-indole-3-carboxaldehyde, 7-benzyloxindole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde, 7-phenylmethoxy
PubChem CID 260799
IUPAC Name 7-phenylmethoxy-1H-indole-3-carbaldehyde
SMILES C1=CC=C(C=C1)COC2=CC=CC3=C2NC=C3C=O

Specifications

Melting Point 152°C to 159°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 251.28
Percent Purity 97%
Chemical Name or Material 7-Benzyloxyindole-3-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.