missing translation for 'onlineSavingsMsg'
Learn More

7-Azaindole, 98%

Product Code. 11322137
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322137 1 g 1g
11332137 5 g 5g
11342137 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11322137 Supplier Thermo Scientific Alfa Aesar Supplier No. L07983.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 271-63-6 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 271-63-6
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005606,MFCD00075874
InChI Key MVXVYAKCVDQRLW-UHFFFAOYSA-N
Synonym 7-azaindole, 1h-pyrrolo 2,3-b pyridine, 1,7-diazaindene, 7h-pyrrolo 2,3-b pyridine, 7-aza-1-pyrindine, 1,7-dideazapurine, 1 h-pyrrolo 2,3:b pyridine, 7-aza indole, unii-qx4465nr9t
PubChem CID 9222
IUPAC Name 1H-pyrrolo[2,3-b]pyridine
SMILES N1C=CC2=CC=CN=C12

Specifications

Melting Point 103°C to 107°C
Boiling Point 270°C
Quantity 1 g
Beilstein 109667
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 7-Azaindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.