missing translation for 'onlineSavingsMsg'
Learn More

7-Azaindole, 98%

Product Code. 10077962 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10077962 1 g Glass bottle 1g
10539620 5 g Glass bottle 5g
11976591 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10077962 Supplier Thermo Scientific Acros Supplier No. 105100010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 271-63-6 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 271-63-6
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005606,MFCD00075874
InChI Key MVXVYAKCVDQRLW-UHFFFAOYSA-N
Synonym 7-azaindole, 1h-pyrrolo 2,3-b pyridine, 1,7-diazaindene, 7h-pyrrolo 2,3-b pyridine, 7-aza-1-pyrindine, 1,7-dideazapurine, 1 h-pyrrolo 2,3:b pyridine, 7-aza indole, unii-qx4465nr9t
PubChem CID 9222
IUPAC Name 1H-pyrrolo[2,3-b]pyridine
SMILES N1C=CC2=CC=CN=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 103.0°C to 107.0°C
Color White
Boiling Point 270°C (1004.0 HPA)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 23, V,6, 306
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, acetonitrile
Formula Weight 118.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 7-Azaindole
Show More Show Less
compliance-icons
Product Identifier
  • 7-Azaindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.