missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Aminothieno[2,3-b]pyrazine-6-carboxylic acid, ≥90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 90.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 195.2 |
Percent Purity | 90% |
Chemical Name or Material | 7-Aminothieno[2, 3-b]pyrazine-6-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'