missing translation for 'onlineSavingsMsg'
Learn More

7-Aminoheptanoic acid, 98%

Product Code. 11491780
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491780 1 g 1g
15453707 250 mg 250mg
11401790 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491780 Supplier Thermo Scientific Alfa Aesar Supplier No. H27519.03
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 929-17-9 | C7H15NO2 | 145.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 929-17-9
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.202
MDL Number MFCD00008242
InChI Key XDOLZJYETYVRKV-UHFFFAOYSA-N
Synonym 7-aminoenanthic acid, heptanoic acid, 7-amino, omega-aminoenantic acid, aminoenanthic acid, amino-enanthylic acid, 7-amino-heptanoic acid, 7-aminohepentanoic acid, omega-aminoheptanoic acid, unii-b10hjx2t48, kyselina 7-aminoenanthova czech
PubChem CID 13580
IUPAC Name 7-aminoheptanoic acid
SMILES C(CCCN)CCC(=O)O

Specifications

Melting Point 194°C to 196°C
Quantity 1 g
Beilstein 906887
Formula Weight 145.19
Percent Purity 98%
Chemical Name or Material 7-Aminoheptanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.