missing translation for 'onlineSavingsMsg'
Learn More

7-Aminodesacetoxycephalosporanic Acid, TRC

Product Code. 30003874
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003874 250 mg 250mg
30007882 500 mg 500mg
17983585 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003874 Supplier Toronto Research Chemicals Supplier No. TRCA604330250MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22252-43-3
Molecular Formula C8 H10 N2 O3 S
Molecular Weight (g/mol) 214.24
Synonym 7-Aminodesacetoxycephalosporanic Acid, 7-ADCA, (6R,7R)-7-Amino-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, Cefadroxil Monohydrate Imp. B (EP), Cefradine Imp. A (EP), Cefalexin Monohydrate Imp. B (EP)
IUPAC Name (6R,7R)-7-amino-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC1=C(N2[C@H](SC1)[C@H](N)C2=O)C(=O)O

Specifications

Analyte or Component Names 7-Aminodesacetoxycephalosporanic Acid
Shipping Condition Room Temperature
InChI Formula InChI=1S/C8H10N2O3S/c1-3-2-14-7-4(9)6(11)10(7)5(3)8(12)13/h4,7H,2,9H2,1H3,(H,12,13)/t4-,7-/m1/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 250 mg
Formula Weight 214.0412
Percent Purity >95%
Chemical Name or Material 7-Aminodesacetoxycephalosporanic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.