missing translation for 'onlineSavingsMsg'
Learn More

7-Aminodesacetoxycephalosporanic Acid, Mikromol™

Product Code. 30459958
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459958 4 x 25 mg 100mg
17981996 25 mg 25mg
17961116 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30459958 Supplier Mikromol Supplier No. MM0811.0100254AMP
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

7-Aminodesacetoxycephalosporanic Acid, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Aminodesacetoxycephalosporanic Acid
CAS 22252-43-3
Analyte or Component Names 7-Aminodesacetoxycephalosporanic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H10 N2 O3 S
Certifications/Compliance ISO 9001
Synonym Cefadroxil Monohydrate Imp. B (EP), Cefadroxil Imp. B (EP), Cefalexin Monohydrate Imp. B (EP), Cefalexin Imp. B (EP), Cefradine Imp. A (EP), Cefadroxil USP Related Compound B, Cefadroxil USP RC B, 7-Aminodesacetoxycephalosporanic Acid, 7-ADCA, (6R,7R)-7-Amino-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, 7-Aminodesacetoxycephalosporanic acid, Cephalexin related, Cefadroxil Related Compound B, Cefadroxil Monohydrate Impurity B, Cefalexin Monohydrate Impurity B, Cefradine Impurity A, Cefadroxil Impurity B, Cefalexin Impurity B
InChI Formula InChI=1S/C8H10N2O3S/c1-3-2-14-7-4(9)6(11)10(7)5(3)8(12)13/h4,7H,2,9H2,1H3,(H,12,13)/t4-,7-/m1/s1
Solution Type Neat
SMILES CC1=C(N2[C@H](SC1)[C@H](N)C2=O)C(=O)O
IUPAC Name (6R,7R)-7-amino-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Molecular Weight (g/mol) 214.24
Quantity 4 x 25 mg
Formula Weight 214.0412 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.