missing translation for 'onlineSavingsMsg'
Learn More

7-Aminodesacetoxycephalosporanic acid, 98%, Thermo Scientific™

Product Code. 11342146
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342146 250 mg 250mg
11322146 1 g 1g
11332146 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342146 Supplier Thermo Scientific Alfa Aesar Supplier No. L03417.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
7-Aminodesacetoxycephalosporanic acid is the Cephalexin metabolite. It is a Potent inhibitor of bacterial cell-wall enzyme. It is used in the synthesis of cephalosporins and for bioconversion studies.

Solubility
Soluble in water (partly), 1M ammonium hydroxide (50 mg/ml), and DMSO (Sparingly).

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22252-43-3
Molecular Formula C8H10N2O3S
Molecular Weight (g/mol) 214.239
MDL Number MFCD00151456
InChI Key NVIAYEIXYQCDAN-CLZZGJSISA-N
Synonym 7-aminodesacetoxycephalosporanic acid, 6r,7r-7-amino-3-methyl-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid, 7-aminodeacetoxycephalosporanic acid, 7-adca, unii-anm3msm8tn, anm3msm8tn, 7beta-amino-3-methyl-3-cephem-4-carboxylic acid, 7beta-amino-3-methylceph-3-em-4-carboxylic acid, 7-amino desacetoxy cephalosporanic acid, 7beta-aminodeacetoxycephalosporanic acid
PubChem CID 33498
ChEBI CHEBI:64984
IUPAC Name (6R,7R)-7-amino-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC1=C(N2C(C(C2=O)N)SC1)C(=O)O

Specifications

Melting Point ∽234°C (decomposition)
Quantity 250 mg
Beilstein 5273616
Solubility Information Soluble in water (partly),1M ammonium hydroxide (50mg/ml),and DMSO (Sparingly).
Formula Weight 214.24
Percent Purity 98%
Chemical Name or Material 7-Aminodesacetoxycephalosporanic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.