missing translation for 'onlineSavingsMsg'
Learn More

7-Aminocephalosporanic acid, MedChemExpress

Product Code. 16498756
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498756 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16498756 Supplier Medchem Express Supplier No. HYB1434
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 7-Aminocephalosporanic acid is the core chemical structure for the synthesis of cephalosporin antibiotics, is a potent β-lactamase inhibitor.

7-Aminocephalosporanic acid is the core chemical structure for the synthesis of cephalosporin antibiotics, is a potent β-lactamase inhibitor.

Specifications

Chemical Name or Material 7-Aminocephalosporanic acid
Physical Form Solid
CAS 957-68-6
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H12N2O5S
For Use With (Application) COVID-19-immunoregulation
Synonym 7-ACA
Solubility Information H2O : 1.1 mg/mL (4.04 mM; ultrasonic and adjust pH to 14 with NaOH) ∣Ethanol : 1.1 mg/mL (4.04 mM; Need ultrasonic) ∣DMF : < 1 mg/mL (insoluble) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=C(C(N12)=C(COC(C)=O)CS[C@]2([H])[C@H](N)C1=O)O
Molecular Weight (g/mol) 272.28
Formula Weight 272.28
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.