missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-4-(trifluoromethyl)coumarin, 97%

Product Code. p-7024154
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
This item is not returnable. View return policy

Product Code. 11408337

Brand: Thermo Scientific Alfa Aesar A18400.01

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53518-15-3 | C10H6F3NO2 | 229.158 g/mol

7-Amino-4-(trifluoromethyl)coumarin is used in the synthesis of substrates for the fluorometric assay of proteolytic enzymes in biological fluids. It is suitable as laser dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Amino-4-(trifluoromethyl)coumarin is used in the synthesis of substrates for the fluorometric assay of proteolytic enzymes in biological fluids. It is suitable as laser dye.

Solubility
Soluble in dimethyl sulfoxide (25 mg/ml), dimethyl formamide (25 mg/ml), and methanol (25 mg/ml). Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53518-15-3
Molecular Formula C10H6F3NO2
Molecular Weight (g/mol) 229.158
MDL Number MFCD00006858
InChI Key JBNOVHJXQSHGRL-UHFFFAOYSA-N
Synonym coumarin 151, 7-amino-4-trifluoromethyl coumarin, 7-amino-4-trifluoromethylcoumarin, 7-amino-4-trifluoromethyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-trifluoromethyl, 7-amino-4-trifluoromethyl chromen-2-one, 7-amino-4-trifluoromethyl-2-benzopyrone, 7-amino-4-trifluoromethyl-chromen-2-one, pubchem8665, maybridge1_007017
PubChem CID 100641
ChEBI CHEBI:51772
IUPAC Name 7-amino-4-(trifluoromethyl)chromen-2-one
SMILES C1=CC2=C(C=C1N)OC(=O)C=C2C(F)(F)F

Specifications

Melting Point 222°C to 223°C
Quantity 0.25 g
Beilstein 4456797
Solubility Information Soluble in dimethyl sulfoxide (25mg/ml),dimethyl formamide (25mg/ml),and methanol (25mg/ml). Insoluble in water.
Formula Weight 229.16
Percent Purity 99%
Chemical Name or Material 7-Amino-4-(trifluoromethyl)coumarin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.