missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 7-Amino-4-(trifluoromethyl)coumarin

Product Code. 16418926
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418926 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418926 Supplier MedChemExpress Supplier No. HYD0981
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 7-Amino-4-(trifluoromethyl)coumarin (Coumarin 151) is a fluorescent marker for the sensitive detection of proteinases. The excitation and emission wavelengths are 400 and 490 nm, respectively.

Specifications

CAS 53518-15-3
Solubility DMSO : 100 mg/mL (436.38 mM∣Need ultrasonic)
Synonym Coumarin 151∣ AFC
Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Quantity 100 mg
Molecular Weight (g/mol) 229.16 g/mol
Product Type Dye Reagents
Purity or Quality Grade 98.92%
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.