missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-4-methylcoumarin, 98%

Product Code. 11487257
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487257 5 g 5g
11384629 250 mg 250mg
11477257 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487257 Supplier Thermo Scientific Alfa Aesar Supplier No. A15017.06
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26093-31-2 | C10H9NO2 | 175.19 g/mol

Used as matrix for analysis of monosulfated disaccharides and as a co-matrix with 6-aza-2-thiothymidine for sulfated neutral and salivated tri-and tetrasaccharides. Reagent for preparing new fluorogenic AMC-based substrates for cystine amino peptidase (and other hydrolases). Also used as reference compound in the enzyme assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as matrix for analysis of monosulfated disaccharides and as a co-matrix with 6-aza-2-thiothymidine for sulfated neutral and salivated tri-and tetrasaccharides. Reagent for preparing new fluorogenic AMC-based substrates for cystine amino peptidase (and other hydrolases). Also used as reference compound in the enzyme assay.

Solubility
Soluble in acetone (10 mg/ml) dimethyl sulfoxide (10 mg/ml) and dimethylformamide.

Notes
Light Sensitive, store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26093-31-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00006868
InChI Key GLNDAGDHSLMOKX-UHFFFAOYSA-N
Synonym 7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x
PubChem CID 92249
ChEBI CHEBI:51771
IUPAC Name 7-amino-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=CC(N)=CC=C12

Specifications

Melting Point 225°C to 228°C
Quantity 5 g
Beilstein 142231
Solubility Information Soluble in acetone (10mg/ml) dimethyl sulfoxide (10mg/ml) and dimethylformamide.
Formula Weight 175.19
Percent Purity 98%
Chemical Name or Material 7-Amino-4-methylcoumarin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.