missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-3,4-dihydro-1(2H)-naphthalenone, 97%

Product Code. p-4228309 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10098329 1 g Glass bottle 1g
10250070 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10098329 Supplier Thermo Scientific Acros Supplier No. 433260010
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22009-40-1 | C10H11NO | 161.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22009-40-1
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD07369817
InChI Key IDNLVJHOEZJNHW-UHFFFAOYSA-N
Synonym 7-amino-3,4-dihydronaphthalen-1 2h-one, 7-amino-1-tetralone, 7-amino-alpha-tetralone, 7-amino-3,4-dihydro-1 2h-naphthalenone, 1 2h-naphthalenone, 7-amino-3,4-dihydro, 7-amino-3,4-dihydronaphthalen-2 1h-one, 7-amino-1,2,3,4-tetrahydronaphthalen-1-one, 7-amino-2,3,4-trihydronaphthalen-1-one, 7-amino-1-oxo-1,2,3,4-tetrahydronaphthalene, 7-aminotetralone
PubChem CID 327177
IUPAC Name 7-amino-3,4-dihydro-2H-naphthalen-1-one
SMILES NC1=CC=C2CCCC(=O)C2=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 138°C to 140°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 161.2
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 7-Amino-3, 4-dihydro-1(2H)-naphthalenone
Show More Show Less
compliance-icons
Product Identifier
  • 7-Amino-3,4-dihydro-1(2H)-naphthalenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.