missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-1H-indazole, 95%, Thermo Scientific™

Product Code. 10431138 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10431138 5 g 5g
10760910 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10431138 Supplier Thermo Scientific Acros Supplier No. 429390050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 21443-96-9
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00022790
InChI Key OTFFCAGPSWJBDK-UHFFFAOYSA-N
Synonym 7-amino-1h-indazole, 2h-indazol-7-amine, 7-aminoindazole, 1h-indazol-7-ylamine, 7-aminindazole, 1h-indazole-7-ylamine, pubchem20891, 1h-indazol-7-amine, 1, cbi-bb zero/005543
PubChem CID 88901
IUPAC Name 1H-indazol-7-amine
SMILES C1=CC2=C(C(=C1)N)NN=C2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 164.0°C to 164.5°C
Color Tan
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 133.15
Percent Purity 95%
Physical Form Crystalline Solid
Chemical Name or Material 7-Amino-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 7-Amino-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.