missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7,9-Dioxo-8-azaspiro[4.5]decane-8-butanoic Acid, Mikromol™
Description
7,9-Dioxo-8-azaspiro[4.5]decane-8-butanoic Acid
Specifications
Specifications
| Chemical Name or Material | 7,9-Dioxo-8-azaspiro[4.5]decane-8-butanoic Acid |
| CAS | 105920-65-8 |
| Analyte or Component Names | 7,9-Dioxo-8-azaspiro[4.5]decane-8-butanoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H19 N O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 7,9-Dioxo-8-azaspiro[4.5]decane-8-butanoic Acid |
| InChI Formula | InChI=1S/C13H19NO4/c15-10-8-13(5-1-2-6-13)9-11(16)14(10)7-3-4-12(17)18/h1-9H2,(H,17,18) |
| SMILES | OC(=O)CCCN1C(=O)CC2(CCCC2)CC1=O |
| IUPAC Name | 4-(7,9-dioxo-8-azaspiro[4.5]decan-8-yl)butanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?