missing translation for 'onlineSavingsMsg'
Learn More

7,8-Dimethoxy-1,3-dihydro-2H-3-benzazepin-2-one, TRC

Product Code. 30383571
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383571 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383571 Supplier Toronto Research Chemicals Supplier No. TRCD461010250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7,8-Dimethoxy-1,3-dihydro-2H-3-benzazepin-2-one
CAS 73942-87-7
Molecular Formula C12 H13 N O3
Quantity 250 mg
Synonym 2H-3-Benzazepin-2-one, 1,3-dihydro-7,8-dimethoxy-, 1,3-Dihydro-7,8-dimethoxy-2H-3-benzazepin-2-one, 7,8-Dimethoxy-1,2-dihydro-3H-3-benzazepin-2-one
InChI Formula InChI=1S/C12H13NO3/c1-15-10-5-8-3-4-13-12(14)7-9(8)6-11(10)16-2/h3-6H,7H2,1-2H3,(H,13,14)
SMILES COc1cc2CC(=O)NC=Cc2cc1OC
IUPAC Name 7,8-dimethoxy-1,3-dihydro-3-benzazepin-2-one
Molecular Weight (g/mol) 219.24
Formula Weight 219.0895
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.