missing translation for 'onlineSavingsMsg'
Learn More

7,8-Dihydroxyflavone, MedChemExpress

Product Code. 16415457
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16415457 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16415457 Supplier Medchem Express Supplier No. HYW013372
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 7,8-Dihydroxyflavone is a potent and selective TrkB agonist that mimics the physiological actions of Brain-derived neurotrophic factor (BDNF). Displays therapeutic efficacy toward various neurological diseases.

7,8-Dihydroxyflavone is a potent and selective TrkB agonist that mimics the physiological actions of Brain-derived neurotrophic factor (BDNF). Displays therapeutic efficacy toward various neurological diseases.

Specifications

Chemical Name or Material 7,8-Dihydroxyflavone
CAS 38183-03-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10O4
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (393.33 mM)
SMILES O=C1C=C(C2=CC=CC=C2)OC3=C(O)C(O)=CC=C13
Molecular Weight (g/mol) 254.24
Formula Weight 254.24
Percent Purity 99.01%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.