missing translation for 'onlineSavingsMsg'
Learn More

7,7-Dichlorobicyclo[3.2.0]hept-2-en-6-one, 97%, Thermo Scientific™

Product Code. 11429036
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429036 5 g 5g
11439036 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429036 Supplier Thermo Scientific Alfa Aesar Supplier No. A17915.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5307-99-3
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00013738
InChI Key JBPBARAOHIDZPU-UHFFFAOYSA-N
Synonym 7,7-dichlorobicyclo 3.2.0 hept-2-en-6-one, 7,7-dichlorobicyclo 3.2.0 hept-2-ene-6-one, bicyclo 3.2.0 hept-2-en-6-one, 7,7-dichloro, 7,7-dichlorobicyclo 3,2,0 hept-2-en-6-one, acmc-20alxm, 6,6-dichlorobicyclo 3.2.0 hept-3-en-7-one, 7,7-dichlorobicyclo-3,2,0 hept-2-en-6-one, 7,7-dichlorobicyclo 3.2.0 hept-2-en-6-one #, bicyclo 3.2.0 hept-2-en-6-one,7,7-dichloro
PubChem CID 98378
IUPAC Name 6,6-dichlorobicyclo[3.2.0]hept-3-en-7-one
SMILES C1C=CC2C1C(=O)C2(Cl)Cl

Specifications

Boiling Point 127°C to 128°C (25 mmHg)
Refractive Index 1.515
Quantity 5 g
Beilstein 2045059
Formula Weight 177.03
Percent Purity 97%
Chemical Name or Material 7,7-Dichlorobicyclo[3.2.0]hept-2-en-6-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.