missing translation for 'onlineSavingsMsg'
Learn More

7,7,8,8-Tetracyanoquinodimethane, 98%

Product Code. 10503324 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10503324 1 g Glass bottle 1g
10657142 5 g Glass bottle 5g
10636762 10 g Glass bottle 10g
10668894 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10503324 Supplier Thermo Scientific Acros Supplier No. 169360010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1518-16-7 | C12H4N4 | 204.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1518-16-7
Molecular Formula C12H4N4
Molecular Weight (g/mol) 204.19
MDL Number MFCD00011664
InChI Key PCCVSPMFGIFTHU-UHFFFAOYSA-N
Synonym 7,7,8,8-tetracyanoquinodimethane, tcnq, tetracyanoquinodimethane, tetracyanoquinodimethan, 2,2'-cyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tetracyano-p-quinodimethane, tetracyanoquinodimethan e, quinodimethan, tetracyano, propanedinitrile, 2,2'-2,5-cyclohexadiene-1,4-diylidene bis, 7,7,8,8-tetracyano-p-quinodimethane
PubChem CID 73697
ChEBI CHEBI:52445
IUPAC Name 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile
SMILES C1=CC(=C(C#N)C#N)C=CC1=C(C#N)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 287°C to 289°C
Color Brown-Orange to Green
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Fieser 01,1136; 12,464
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetomitrile,tetrahydrofuran and,dioxane
Formula Weight 204.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 7,7,8,8-Tetracyanoquinodimethane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 7,7,8,8-Tetracyanoquinodimethane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.