missing translation for 'onlineSavingsMsg'
Learn More

7,4'-Di-O-methylapigenin, MedChemExpress

Product Code. 16490386
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490386 2 mg 2mg
16481678 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16490386 Supplier Medchem Express Supplier No. HYN2144

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress The compound 7,4'-Di-O-methylapigenin may be partly responsible for the reported antifungal activity of C.

The compound 7,4'-Di-O-methylapigenin may be partly responsible for the reported antifungal activity of C.

Specifications

Chemical Name or Material 7,4'-Di-O-methylapigenin
Physical Form Solid
CAS 5128-44-9
Color Yellow
Quantity 2 mg
Purity Grade Notes Research
Molecular Formula C17H14O5
For Use With (Application) Neuroscience-Neuromodulation
Synonym 4',7-Dimethoxy-5-Hydroxyflavone
Solubility Information DMSO : 5 mg/mL (16.76 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming) (insoluble)
SMILES O=C1C=C(C2=CC=C(OC)C=C2)OC3=CC(OC)=CC(O)=C13
Molecular Weight (g/mol) 298.29
Formula Weight 298.29
Percent Purity 99.09%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.