missing translation for 'onlineSavingsMsg'
Learn More

7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan, Thermo Scientific™

Product Code. 10302285
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10302285 5 g Amber Glass Bottle 5g
10774847 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10302285 Supplier Thermo Scientific Maybridge Supplier No. CC78239DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1192755-14-8
Molecular Formula C14H17BO3
Molecular Weight (g/mol) 244.10
MDL Number MFCD13810042
InChI Key DNZVVIZYTCIYNY-UHFFFAOYSA-N
Synonym benzofuran-7-boronic acid pinacol ester, 2-benzofuran-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzo b furan, 2-1-benzofuran-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzofuran, benzo b furan-7-boronic acid pinacol ester, 7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-benzofuran, 2-benzo 2,3-b furan-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 45790048
IUPAC Name 2-(1-benzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=C2OC=CC2=CC=C1

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 244.1
Percent Purity 97%
Chemical Name or Material 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.