missing translation for 'onlineSavingsMsg'
Learn More

7,3'-bis(O-(2,3,4-Tri-O-acetyl-(Methyl Beta-D-Glucopyranosiduronate) rac-Hesperetin, TRC

Product Code. 30394525
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394525 5 mg 5mg
30405202 2.5 mg 2.5mg
30394992 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394525 Supplier Toronto Research Chemicals Supplier No. TRCH9727985MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7,3'-bis(O-(2,3,4-Tri-O-acetyl)-(Methyl beta-D-Glucopyranosiduronate)) rac-Hesperetin
Analyte or Component Names 7,3'-bis(O-(2,3,4-Tri-O-acetyl)-(Methyl beta-D-Glucopyranosiduronate)) rac-Hesperetin
Shipping Condition Room Temperature
Molecular Formula C42H46O24
Synonym (2S,3R,4R,5S,6S)-2-((5-hydroxy-2-(4-methoxy-3-(((2R,3S,5R,6R)-3,4,5-triacetoxy-6-(methoxycarbonyl)tetrahydro-2H-pyran-2-yl)oxy)phenyl)-4-oxochroman-7-yl)oxy)-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate
InChI Formula InChI=1S/C42H46O24/c1-16(43)56-31-33(58-18(3)45)37(60-20(5)47)41(65-35(31)39(51)54-8)62-23-13-24(49)30-25(50)15-27(63-29(30)14-23)22-10-11-26(53-7)28(12-22)64-42-38(61-21(6)48)34(59-19(4)46)32(57-17(2)44)36(66-42)40(52)55-9/h10-14,27,31-38,41-42,49H,15H2,1-9H3/t27?,31-,32+,33+,34?,35-,36+,37+,38-,41+,42-/m0/s1
Solution Type Neat
SMILES O=C1C2=C(O)C=C(O[C@@H]3O[C@H](C(OC)=O)[C@@H](OC(C)=O)[C@@H](OC(C)=O)[C@H]3OC(C)=O)C=C2OC(C4=CC=C(OC)C(O[C@H]5O[C@@H](C(OC)=O)[C@H](OC(C)=O)C(OC(C)=O)[C@@H]5OC(C)=O)=C4)C1
Industry Type Pharmaceutical
Molecular Weight (g/mol) 934.8
Quantity 5 mg
Formula Weight 934.24
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.